Tisk…

Smoothened agonist

Dodavatel: Apollo Scientific
Varování

OR958890-1MG OR958890-25MG OR958890-5MG
APOSOR958890-1MGEA 4300 CZK
APOSOR958890-1MG APOSOR958890-25MG APOSOR958890-5MG
Smoothened agonist
Smoothened agonist
Vzorec: C₂₈H₂₈ClN₃OS
Storage Temperature: Freezer
MDL Number: MFCD22581513
CAS číslo: 912545-86-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR