cis-1,3-Cyclohexanedicarboxylic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
cis-Cyclohexane-1,3-dicarboxylic acid
OR958474-10G
OR958474-1G
OR958474-5G
APOSOR958474-10GEA
5630
CZK
APOSOR958474-10G
APOSOR958474-1G
APOSOR958474-5G
cis-1,3-Cyclohexanedicarboxylic acid 95%
cis-1,3-Cyclohexanedicarboxylic acid
Vzorec:
C₈H₁₂O₄ MW: 172,1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134411 CAS číslo: 2305-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...