Tisk…

3,3'-Bipiperidine 95%

Dodavatel: Apollo Scientific

OR958208-1G OR958208-250MG
APOSOR958208-1GEA 14880 CZK
APOSOR958208-1G APOSOR958208-250MG
3,3'-Bipiperidine 95%
3,3'-Bipiperidine
Vzorec: C₁₀H₂₀N₂
MW: 168,28 g/mol
Storage Temperature: Ambient
CAS číslo: 46040-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR