2,3,4-Trichlorophenol 98%
Dodavatel: Apollo Scientific
OR957661-1G
OR957661-250MG
OR957661-5G
APOSOR957661-1GEA
15150
CZK
APOSOR957661-1G
APOSOR957661-250MG
APOSOR957661-5G
2,3,4-Trichlorophenol 98%
2,3,4-Trichlorophenol
Vzorec:
C₆H₃Cl₃O MW: 197.45 Bod varu: 256.8 Storage Temperature: Ambient |
CAS číslo:
15950-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...