4-Chloro-3-nitrophenylboronic acid pinacol ester 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloro-3-nitrobenzeneboronic acid pinacol ester, 4-Chloro-3-nitrophenylboronic acid, pinacol ester, 2-(4-Chloro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR9576-1G
OR9576-5G
APOSOR9576-1GEA
2030
CZK
APOSOR9576-1G
APOSOR9576-5G
4-Chloro-3-nitrophenylboronic acid pinacol ester 98%
4-Chloro-3-nitrophenylboronic acid pinacol ester
Vzorec:
C₁₂H₁₅BClNO₄ MW: 283,52 g/mol Bod tání: 56…58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08235086 CAS číslo: 913836-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...