N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine 95%
Dodavatel: Apollo Scientific
Synonyms:
6-Chloro-N,N-dimethylpyrazin-2-amine
OR957476-25G
APOSOR957476-25GEA
5060
CZK
APOSOR957476-25G
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine 95%
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine
Vzorec:
C₆H₈ClN₃ MW: 157,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12024889 CAS číslo: 61655-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...