Tisk…

3-Isopropylphenol 95%

Dodavatel: Apollo Scientific

OR957472-10G OR957472-1G OR957472-5G
APOSOR957472-10GEA 10350 CZK
APOSOR957472-10G APOSOR957472-1G APOSOR957472-5G
3-Isopropylphenol 95%
3-Isopropylphenol
Vzorec: C₉H₁₂O
MW: 136,19 g/mol
Storage Temperature: Ambient
CAS číslo: 618-45-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR