Ethyl-5-chloronicotinate 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 3-chloro-5-pyridinecarboxylate
OR957081-1G
OR957081-5G
APOSOR957081-1GEA
2070
CZK
APOSOR957081-1G
APOSOR957081-5G
Ethyl-5-chloronicotinate 95%
Ethyl-5-chloronicotinate
Vzorec:
C₈H₈ClNO₂ MW: 185,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01692171 CAS číslo: 20825-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...