Tisk…

2-Chloro-N,N-dimethylaniline 95%

Dodavatel: Apollo Scientific

OR956951-25G OR956951-5G
APOSOR956951-25GEA 4800 CZK
APOSOR956951-25G APOSOR956951-5G
2-Chloro-N,N-dimethylaniline 95%
2-Chloro-N,N-dimethylaniline
Vzorec: C₈H₁₀ClN
Storage Temperature: Ambient
MDL Number: MFCD03452792
CAS číslo: 698-01-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR