Methyl-4-iodopicolinate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 4-iodopyridine-2-carboxylate
OR9563-1G
OR9563-5G
APOSOR9563-1GEA
4530
CZK
APOSOR9563-1G
APOSOR9563-5G
Methyl-4-iodopicolinate 98%
Methyl-4-iodopicolinate
Vzorec:
C₇H₆INO₂ Bod tání: 73…76 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08235100 CAS číslo: 380381-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...