Tisk…

2-Methyl-3-nitrobenzonitrile 97%

Dodavatel: Apollo Scientific

OR956279-1G OR956279-5G
APOSOR956279-1GEA 612 CZK
APOSOR956279-1G APOSOR956279-5G
2-Methyl-3-nitrobenzonitrile 97%
2-Methyl-3-nitrobenzonitrile
Vzorec: C₈H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09025714
CAS číslo: 71516-35-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR