Tisk…

{4-[(Methylsulphonyl)methyl]phenyl}amine hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

OR956229-1G OR956229-250MG
APOSOR956229-1GEA 6290 CZK
APOSOR956229-1G APOSOR956229-250MG
{4-[(Methylsulphonyl)methyl]phenyl}amine hydrochloride 97%
{4-[(Methylsulphonyl)methyl]phenyl}amine hydrochloride
Vzorec: C₈H₁₂ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD18071261
CAS číslo: 24176-70-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR