Tisk…
Methyl-N-acetyl-L-valinate 95%
Dodavatel: Apollo Scientific
Synonyms: (S)-Methyl 2-acetamido-3-methylbutanoate, Ac-Val-OMe
OR955549-1G OR955549-5G
APOSOR955549-1GEA 2440 CZK
APOSOR955549-1G APOSOR955549-5G
Methyl-N-acetyl-L-valinate 95%
Methyl-N-acetyl-L-valinate
Vzorec: C₈H₁₅NO₃
MW: 173,212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080847
CAS číslo: 1492-15-5