Tisk…

4-(2-Chloroethylcarbamoyl)phenylboronic acid 95%

Dodavatel: Apollo Scientific

OR9551-1G OR9551-5G
APOSOR9551-1GEA 3340 CZK
APOSOR9551-1G APOSOR9551-5G
4-(2-Chloroethylcarbamoyl)phenylboronic acid 95%
4-(2-Chloroethylcarbamoyl)phenylboronic acid
Vzorec: C₉H₁₁BClNO₃
Bod tání: 173…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436019
CAS číslo: 874460-05-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR