Tisk…

N-(2-Cyanoethyl)-N-ethyl-m-toluidine 95%

Dodavatel: Apollo Scientific

Synonyms: 3-(N-Ethyl-m-toluidino)propionitrile, 3-[Ethyl(3-methylphenyl)amino]propionitrile

OR954965-1G OR954965-5G
APOSOR954965-1GEA 4770 CZK
APOSOR954965-1G APOSOR954965-5G
N-(2-Cyanoethyl)-N-ethyl-m-toluidine 95%
N-(2-Cyanoethyl)-N-ethyl-m-toluidine
Vzorec: C₁₂H₁₆N₂
MW: 188,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019859
CAS číslo: 148-69-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR