Bifenyl-4-ol 98%
Dodavatel: Apollo Scientific
Synonyms:
[1,1'-Biphenyl]-4-ol, 4-Hydroxydiphenyl, 4-Phenylphenol, p-Hydroxybiphenyl, 4-Hydroxybiphenyl
OR954761-100G
OR954761-500G
APOSOR954761-100GEA
0
CZK
APOSOR954761-100G
APOSOR954761-500G
Bifenyl-4-ol 98%
Bifenyl-4-ol
Vzorec:
C₁₂H₁₀O MW: 170.21 Bod varu: 321 Bod tání: 162 Storage Temperature: Ambient |
MDL Number:
MFCD00002347 CAS číslo: 92-69-3 Merck Index: 12,07459 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...