Tisk…

3-(2-Chloroethylcarbamoyl)phenylboronic acid 96%

Dodavatel: Apollo Scientific

Synonyms: (3-((2-Chloroethyl)carbamoyl)phenyl)boronic acid

OR9545-1G OR9545-5G
APOSOR9545-1GEA 3340 CZK
APOSOR9545-1G APOSOR9545-5G
3-(2-Chloroethylcarbamoyl)phenylboronic acid 96%
3-(2-Chloroethylcarbamoyl)phenylboronic acid
Vzorec: C₉H₁₁BClNO₃
MW: 227,4500 g/mol
Bod tání: 250…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436013
CAS číslo: 874288-12-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR