3-(Benzyloxy)-4-bromobenzoic acid 98%
Dodavatel: Apollo Scientific
OR9541-1G
OR9541-5G
APOSOR9541-1GEA
6550
CZK
APOSOR9541-1G
APOSOR9541-5G
3-(Benzyloxy)-4-bromobenzoic acid 98%
3-(Benzyloxy)-4-bromobenzoic acid
Vzorec:
C₁₄H₁₁BrO₃ Bod tání: 198…201 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08436069 CAS číslo: 17054-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...