Tisk…

4-Methoxymethylbenzoic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(Methoxymethyl)benzoic acid

OR954026-1G OR954026-250MG
APOSOR954026-1GEA 13860 CZK
APOSOR954026-1G APOSOR954026-250MG
4-Methoxymethylbenzoic acid 95%
4-Methoxymethylbenzoic acid
Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06375934
CAS číslo: 67003-50-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR