Tisk…

4,6-Dimethylbenzothiazol-2-ylamine 98%

Dodavatel: Apollo Scientific

OR953987-10G OR953987-1G OR953987-5G
APOSOR953987-10GEA 18420 CZK
APOSOR953987-10G APOSOR953987-1G APOSOR953987-5G
4,6-Dimethylbenzothiazol-2-ylamine 98%
4,6-Dimethylbenzothiazol-2-ylamine
Vzorec: C₉H₁₀N₂S
MW: 178,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00659246
CAS číslo: 64036-71-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR