1-Cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR953972-1G
OR953972-5G
APOSOR953972-1GEA
5790
CZK
APOSOR953972-1G
APOSOR953972-5G
1-Cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde 95%
1-Cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde
Vzorec:
C₁₀H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD04309657 CAS číslo: 640264-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...