3-Amino-N,N-dimethylbenzamide 95%
Dodavatel: Apollo Scientific
OR953934-1G
OR953934-5G
APOSOR953934-1GEA
2440
CZK
APOSOR953934-1G
APOSOR953934-5G
3-Amino-N,N-dimethylbenzamide 95%
3-Amino-N,N-dimethylbenzamide
Vzorec:
C₉H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD01168905 CAS číslo: 33322-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...