Tisk…

2-Piperidin-3-yl-1H-benzimidazole ≥98%

Dodavatel: Apollo Scientific

OR953882-1G OR953882-5G
APOSOR953882-1GEA 1700 CZK
APOSOR953882-1G APOSOR953882-5G
2-Piperidin-3-yl-1H-benzimidazole ≥98%
2-Piperidin-3-yl-1H-benzimidazole
Vzorec: C₁₂H₁₅N₃
Storage Temperature: Ambient
MDL Number: MFCD06738924
CAS číslo: 123771-23-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR