5-Benzylthiazol-2-ylamine 95%
Dodavatel: Apollo Scientific
OR953871-1G
OR953871-5G
APOSOR953871-1GEA
6420
CZK
APOSOR953871-1G
APOSOR953871-5G
5-Benzylthiazol-2-ylamine 95%
5-Benzylthiazol-2-ylamine
Vzorec:
C₁₀H₁₀N₂S MW: 190,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00547831 CAS číslo: 121952-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...