3-(Methanesulphonylamino)benzonitrile 95%
Dodavatel: Apollo Scientific
OR953833-1G
OR953833-5G
APOSOR953833-1GEA
4770
CZK
APOSOR953833-1G
APOSOR953833-5G
3-(Methanesulphonylamino)benzonitrile 95%
3-(Methanesulphonylamino)benzonitrile
Vzorec:
C₈H₈N₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD00116199 CAS číslo: 50790-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...