Tisk…

1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 98%

Dodavatel: Apollo Scientific

OR953815-5G
APOSOR953815-5GEA 6120 CZK
APOSOR953815-5G
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 98%
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile
Vzorec: C₈H₅N₃
MW: 143,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659684
CAS číslo: 517918-95-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR