Tisk…

Isoquinoline-1-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR953813-1G OR953813-250MG
APOSOR953813-1GEA 2480 CZK
APOSOR953813-1G APOSOR953813-250MG
Isoquinoline-1-carbaldehyde 95%
Isoquinoline-1-carbaldehyde
Vzorec: C₁₀H₇NO
MW: 157,1687 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738864
CAS číslo: 4494-18-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR