2-Chloro-4-iodo-5-methylpyridine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR953758-250MG
OR953758-1G
APOSOR953758-250MGEA
1080
CZK
APOSOR953758-250MG
APOSOR953758-1G
2-Chloro-4-iodo-5-methylpyridine 95%
2-Chloro-4-iodo-5-methylpyridine
Vzorec:
C₆H₅ClIN MW: 253.47 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16610529 CAS číslo: 1197957-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...