Tisk…

3-Amino-4-methylpyridazine 95%

Dodavatel: Apollo Scientific

OR953732-1G OR953732-250MG
APOSOR953732-1GEA 16500 CZK
APOSOR953732-1G APOSOR953732-250MG
3-Amino-4-methylpyridazine 95%
3-Amino-4-methylpyridazine
Vzorec: C₅H₇N₃
MW: 109.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460975
CAS číslo: 90568-15-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR