Tisk…

3,3,5-Trimethylcyclohexanol 98%

Dodavatel: Apollo Scientific

OR953657-100G OR953657-10G OR953657-25G
APOSOR953657-100GEA 2570 CZK
APOSOR953657-100G APOSOR953657-10G APOSOR953657-25G
3,3,5-Trimethylcyclohexanol 98%
3,3,5-Trimethylcyclohexanol
Vzorec: C₉H₁₈O
MW: 142,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019378
CAS číslo: 116-02-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR