Tisk…

9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 9-Benzyl-9-azabicyclo[3.3.1]nonan-3-one

OR953603-5G
APOSOR953603-5GEA 2360 CZK
APOSOR953603-5G
9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane 97%
9-Benzyl-3-oxo-9-azabicyclo[3.3.1]nonane
Vzorec: C₁₅H₁₉NO
MW: 229,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144851
CAS číslo: 2291-58-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR