Ethyl-1-methylpyrrolidine-3-carboxylate 95%
Dodavatel: Apollo Scientific
OR953602-1G
OR953602-250MG
APOSOR953602-1GEA
7080
CZK
APOSOR953602-1G
APOSOR953602-250MG
Ethyl-1-methylpyrrolidine-3-carboxylate 95%
Ethyl-1-methylpyrrolidine-3-carboxylate
Vzorec:
C₈H₁₅NO₂ MW: 157,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12198863 CAS číslo: 14398-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...