Tisk…

2-Cyano-N-(2-phenylethyl)acetamide 95%

Dodavatel: Apollo Scientific

OR953526-1G OR953526-5G
APOSOR953526-1GEA 14290 CZK
APOSOR953526-1G APOSOR953526-5G
2-Cyano-N-(2-phenylethyl)acetamide 95%
2-Cyano-N-(2-phenylethyl)acetamide
Vzorec: C₁₁H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00552868
CAS číslo: 51838-02-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR