5-Ethyl-4-methylthiophene-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR953479-100MG
OR953479-1G
OR953479-250MG
APOSOR953479-100MGEA
4770
CZK
APOSOR953479-100MG
APOSOR953479-1G
APOSOR953479-250MG
5-Ethyl-4-methylthiophene-2-carboxylic acid 95%
5-Ethyl-4-methylthiophene-2-carboxylic acid
Vzorec:
C₈H₁₀O₂S Storage Temperature: Ambient |
MDL Number:
MFCD06654975 CAS číslo: 40808-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...