Tisk…

4-Methylisatin 95%

Dodavatel: Apollo Scientific

OR953472-1G OR953472-250MG
APOSOR953472-1GEA 17990 CZK
APOSOR953472-1G APOSOR953472-250MG
4-Methylisatin 95%
4-Methylisatin
Vzorec: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00463404
CAS číslo: 1128-44-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR