Tisk…

2,4-Dichloro-5-pyrimidinecarboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2,4-Dichloropyrimidine-5-carboxylic acid

OR953411-10G OR953411-1G OR953411-5G
APOSOR953411-10GEA 23410 CZK
APOSOR953411-10G APOSOR953411-1G APOSOR953411-5G
2,4-Dichloro-5-pyrimidinecarboxylic acid 95%
2,4-Dichloro-5-pyrimidinecarboxylic acid
Vzorec: C₅H₂Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09999133
CAS číslo: 37131-89-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR