Tisk…

1-(2-Nitrophenyl)ethanol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Nitro-alpha-methylbenzyl alcohol

OR953402-5G
APOSOR953402-5GEA 2610 CZK
APOSOR953402-5G
1-(2-Nitrophenyl)ethanol 95%
1-(2-Nitrophenyl)ethanol
Vzorec: C₈H₉NO₃
MW: 167,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683380
CAS číslo: 3205-25-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR