Methyl-3-methylthiophene-2-carboxylate 96%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 3-methylthiophene-2-carboxylate
OR953363-25G
APOSOR953363-25GEA
2440
CZK
APOSOR953363-25G
Methyl-3-methylthiophene-2-carboxylate 96%
Methyl-3-methylthiophene-2-carboxylate
Vzorec:
C₇H₈O₂S MW: 156,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234332 CAS číslo: 81452-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...