1-(3-Methylphenyl)cyclopropanamine 95%
Dodavatel: Apollo Scientific
Synonyms:
1-(m-Tolyl)cyclopropanamine, 1-(p-tolyl)cyclopropanamine
OR953037-1G
OR953037-250MG
APOSOR953037-1GEA
14720
CZK
APOSOR953037-1G
APOSOR953037-250MG
1-(3-Methylphenyl)cyclopropanamine 95%
1-(3-Methylphenyl)cyclopropanamine
Vzorec:
C₁₀H₁₃N MW: 147,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07374481 CAS číslo: 503417-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...