Tisk…

Boc-D-Alanine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Boc-D-alanine, Boc-D-Ala-OH

OR952941-100G OR952941-1KG OR952941-500G
APOSOR952941-100GEA 2110 CZK
APOSOR952941-100G APOSOR952941-1KG APOSOR952941-500G
Boc-D-Alanine 98%
Boc-D-Alanine
Vzorec: C₈H₁₅NO₄
MW: 189,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063123
CAS číslo: 7764-95-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR