Tisk…

Ethyl tetrahydropyran-4-carboxylate 98%

Dodavatel: Apollo Scientific

Synonyms: Ethyl tetrahydro-2H-pyran-4-carboxylate

OR952913-5G
APOSOR952913-5GEA 2070 CZK
APOSOR952913-5G
Ethyl tetrahydropyran-4-carboxylate 98%
Ethyl tetrahydropyran-4-carboxylate
Vzorec: C₈H₁₄O₃
MW: 158,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08702763
CAS číslo: 96835-17-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR