Tisk…

2-(4-Methylphenoxy)aniline 98%

Dodavatel: Apollo Scientific

OR952890-1G OR952890-5G
APOSOR952890-1GEA 6220 CZK
APOSOR952890-1G APOSOR952890-5G
2-(4-Methylphenoxy)aniline 98%
2-(4-Methylphenoxy)aniline
Vzorec: C₁₃H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00035923
CAS číslo: 20927-98-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR