8-Chloro-3,4-dihydro-2H-naphthalen-1-one 98%
Dodavatel: Apollo Scientific
Synonyms:
8-Chloro-3,4-dihydronaphthalen-1(2H)-one
OR952727-1G
OR952727-250MG
APOSOR952727-1GEA
5960
CZK
APOSOR952727-1G
APOSOR952727-250MG
8-Chloro-3,4-dihydro-2H-naphthalen-1-one 98%
8-Chloro-3,4-dihydro-2H-naphthalen-1-one
Vzorec:
C₁₀H₉ClO MW: 180,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06796577 CAS číslo: 68449-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...