Tisk…

2-Methoxy-2-methylpropanoic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Methoxy-2-methylpropionic Acid

OR952689-1G OR952689-5G OR952689-10G
APOSOR952689-1GEA 2360 CZK
APOSOR952689-1G APOSOR952689-5G APOSOR952689-10G
2-Methoxy-2-methylpropanoic acid 98%
2-Methoxy-2-methylpropanoic acid
Vzorec: C₅H₁₀O₃
MW: 118,13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD07787581
CAS číslo: 13836-62-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR