Tisk…
1,2-Cyclohexanedicarboximide 95%
Dodavatel: Apollo Scientific
Synonyms: 1,2-Cyclohexanedicarboximide, Hexahydrophthalimide
OR952647-100G OR952647-500G
APOSOR952647-100GEA 1120 CZK
APOSOR952647-100G APOSOR952647-500G
1,2-Cyclohexanedicarboximide 95%
1,2-Cyclohexanedicarboximide
Vzorec: C₈H₁₁NO₂
MW: 153.1784 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035737
CAS číslo: 1444-94-6