Tisk…

N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide 95%

Dodavatel: Apollo Scientific

OR952632-1G OR952632-250MG
APOSOR952632-1GEA 12830 CZK
APOSOR952632-1G APOSOR952632-250MG
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide 95%
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide
Vzorec: C₁₀H₁₄N₂O₂
MW: 194.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07776841
CAS číslo: 169205-93-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR