7-Hydroxy-1,2,3,4-tetrahydroquinoline 98%
Dodavatel: Apollo Scientific
OR952616-1G
OR952616-5G
OR952616-10G
APOSOR952616-1GEA
1410
CZK
APOSOR952616-1G
APOSOR952616-5G
APOSOR952616-10G
7-Hydroxy-1,2,3,4-tetrahydroquinoline 98%
7-Hydroxy-1,2,3,4-tetrahydroquinoline
Vzorec:
C₉H₁₁NO MW: 149,1897 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08063887 CAS číslo: 58196-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...