Tisk…

7-Ethyl-1H-indole-2,3-dione 95%

Dodavatel: Apollo Scientific

Synonyms: 7-Ethylisatin

OR952606-1G OR952606-5G
APOSOR952606-1GEA 6020 CZK
APOSOR952606-1G APOSOR952606-5G
7-Ethyl-1H-indole-2,3-dione 95%
7-Ethyl-1H-indole-2,3-dione
Vzorec: C₁₀H₉NO₂
MW: 175,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047222
CAS číslo: 79183-65-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR