7-Bromo-2-chloroquinoxaline 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR952570-1G
OR952570-5G
APOSOR952570-1GEA
622
CZK
APOSOR952570-1G
APOSOR952570-5G
7-Bromo-2-chloroquinoxaline 97%
7-Bromo-2-chloroquinoxaline
Vzorec:
C₈H₄BrClN₂ MW: 243,4900 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02955309 CAS číslo: 89891-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...