N-(4-Hydroxy-pyridin-3-yl)-2,2-dimethylpropionamide 95%
Dodavatel: Apollo Scientific
OR952514-1G
OR952514-250MG
APOSOR952514-1GEA
12830
CZK
APOSOR952514-1G
APOSOR952514-250MG
N-(4-Hydroxy-pyridin-3-yl)-2,2-dimethylpropionamide 95%
N-(4-Hydroxy-pyridin-3-yl)-2,2-dimethylpropionamide
Vzorec:
C₁₀H₁₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08235134 CAS číslo: 540497-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...